Product Details

SNP ID
rs59592860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:21845028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTGGTTTACGGTTTTAATTCA[A/G]GAAACAATATCAAGACAATAGGAAG
Phenotype
MIM: 608182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNIP4 PubMed Links

Gene Details

Gene
KCNIP4
Gene Name
potassium voltage-gated channel interacting protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035003.1 Intron NP_001030175.1
NM_001035004.1 Intron NP_001030176.1
NM_025221.5 Intron NP_079497.2
NM_147181.3 Intron NP_671710.1
NM_147182.3 Intron NP_671711.1
NM_147183.3 Intron NP_671712.1
XM_005248190.4 Intron XP_005248247.1
XM_011513882.2 Intron XP_011512184.1
XM_011513883.2 Intron XP_011512185.1
XM_011513884.2 Intron XP_011512186.1
XM_011513885.2 Intron XP_011512187.1
XM_011513886.2 Intron XP_011512188.1
XM_011513887.2 Intron XP_011512189.1
XM_011513889.2 Intron XP_011512191.1
XM_017008653.1 Intron XP_016864142.1
Gene
KCNIP4-IT1
Gene Name
KCNIP4 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details