Product Details

SNP ID
rs56675911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:63059848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGGAAGGGGAACATCACACAC[C/T]GGGGCCTATTTATTTATTTTTGAAA
Phenotype
MIM: 600415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTPA PubMed Links

Gene Details

Gene
TTPA
Gene Name
alpha tocopherol transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000370.3 2395 UTR 3 NP_000361.1
XM_006716468.3 2395 UTR 3 XP_006716531.1

View Full Product Details