Product Details

SNP ID
rs60889939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18330886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGACCTATTTCCTTTCTGTTCTC[A/G]GGATAGAATAGCCTGTAAAGACCTA
Phenotype
MIM: 189972 MIM: 607521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2H1 PubMed Links

Gene Details

Gene
GTF2H1
Gene Name
general transcription factor IIH subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142307.1 Intron NP_001135779.1
NM_005316.3 Intron NP_005307.1
XM_006718208.3 Intron XP_006718271.1
Gene
HPS5
Gene Name
HPS5, biogenesis of lysosomal organelles complex 2 subunit 2
There are no transcripts associated with this gene.

View Full Product Details