Product Details
- SNP ID
-
rs57351379
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56644879 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTTTTTTAAACACAAACCTAAC[C/T]CATTTACTATGTCTGTTTTCTTAAA
- Phenotype
-
MIM: 156350
MIM: 156349
MIM: 156356
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MT1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143445434] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MT1A
- Gene Name
- metallothionein 1A
There are no transcripts associated with this gene.
- Gene
- MT1B
- Gene Name
- metallothionein 1B
There are no transcripts associated with this gene.
- Gene
- MT1DP
- Gene Name
- metallothionein 1D, pseudogene
There are no transcripts associated with this gene.
- Gene
- MT1JP
- Gene Name
- metallothionein 1J, pseudogene
There are no transcripts associated with this gene.
View Full Product Details