Product Details

SNP ID
rs62089498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3277630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGAACCTGACTTCATATTAAC[A/G]TCTTTGTTCATCTCTATAGAAACTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR3A2 PubMed Links

Gene Details

Gene
OR3A2
Gene Name
olfactory receptor family 3 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002551.3 1657 Intron NP_002542.3
XM_017024697.1 1657 UTR 3 XP_016880186.1
XM_017024698.1 1657 UTR 3 XP_016880187.1

View Full Product Details