Product Details

SNP ID
rs61750951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38252920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTTTCCGGCTTCTCTCCTCTT[C/G]ACTCTCTAACTCCAACAATTCATCA
Phenotype
MIM: 608153
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R14A PubMed Links

Gene Details

Gene
PPP1R14A
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243947.1 508 Intron NP_001230876.1
NM_033256.2 508 Missense Mutation CAA,GAA Q86E NP_150281.1

View Full Product Details