Product Details

SNP ID
rs62119261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44618732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTATCCCAGGCTTGTTCACTC[A/C]GCAATGGAAGAGGAACAAGAGGTAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGSF23 PubMed Links

Gene Details

Gene
IGSF23
Gene Name
immunoglobulin superfamily member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205280.1 Intron NP_001192209.1
XM_017026317.1 Intron XP_016881806.1
XM_017026318.1 Intron XP_016881807.1
XM_017026319.1 Intron XP_016881808.1
XM_017026320.1 Intron XP_016881809.1
XM_017026321.1 Intron XP_016881810.1
XM_017026322.1 Intron XP_016881811.1
XM_017026323.1 Intron XP_016881812.1
XM_017026324.1 Intron XP_016881813.1
XM_017026325.1 Intron XP_016881814.1
XM_017026326.1 Intron XP_016881815.1

View Full Product Details