Product Details

SNP ID
rs61752964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120535887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGGTTGACTGCCACTGAAGTTT[C/T]CTGCCACTATGTTTGCCTAGGTAAA
Phenotype
MIM: 300304
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUL4B PubMed Links

Gene Details

Gene
CUL4B
Gene Name
cullin 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079872.1 2427 Silent Mutation AGA,AGG R701R NP_001073341.1
NM_003588.3 2427 Silent Mutation AGA,AGG R719R NP_003579.3
XM_005262481.1 2427 Silent Mutation AGA,AGG R721R XP_005262538.1
XM_006724784.1 2427 Silent Mutation AGA,AGG R708R XP_006724847.1
XM_006724785.2 2427 Silent Mutation AGA,AGG R706R XP_006724848.1
XM_011531399.2 2427 Silent Mutation AGA,AGG R525R XP_011529701.1
XM_011531400.2 2427 Silent Mutation AGA,AGG R523R XP_011529702.1
XM_011531401.1 2427 Silent Mutation AGA,AGG R507R XP_011529703.1

View Full Product Details