Product Details

SNP ID
rs58540395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85546059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATCTGAATGGTGGTCCAGCTT[A/G]TCCTGCAAATGTAGAGCCAAGGAGT
Phenotype
MIM: 137167 MIM: 601468 MIM: 616350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGCX PubMed Links

Gene Details

Gene
GGCX
Gene Name
gamma-glutamyl carboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000821.6 6258 UTR 3 NP_000812.2
NM_001142269.3 6258 UTR 3 NP_001135741.1
NM_001311312.1 6258 Intron NP_001298241.1
XM_005264259.4 6258 Intron XP_005264316.1
XM_011532764.2 6258 Intron XP_011531066.1
XM_011532765.2 6258 Intron XP_011531067.1
XM_017003803.1 6258 Intron XP_016859292.1
Gene
MAT2A
Gene Name
methionine adenosyltransferase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005911.5 6258 Intron NP_005902.1
Gene
PARTICL
Gene Name
promoter of MAT2A antisense radiation-induced circulating long non-coding RNA
There are no transcripts associated with this gene.

View Full Product Details