Product Details

SNP ID
rs61987160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54513404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGAGAAGTGAGAAGTGAGATA[C/G]GTGCTTCTTTGTTCACTGGAGCACT
Phenotype
MIM: 606138
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CGRRF1 PubMed Links

Gene Details

Gene
CGRRF1
Gene Name
cell growth regulator with ring finger domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006568.2 Intron NP_006559.1
XM_011536362.2 Intron XP_011534664.1
XM_017020931.1 Intron XP_016876420.1
XM_017020932.1 Intron XP_016876421.1

View Full Product Details