Product Details

SNP ID
rs55654701
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:37958570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTCTATGTTTAAGATGGGGAA[A/C]CCTGGTTGAGAAAGGCTGACCTACA
Phenotype
MIM: 605596
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SF3A3 PubMed Links

Gene Details

Gene
SF3A3
Gene Name
splicing factor 3a subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320830.1 Intron NP_001307759.1
NM_006802.3 Intron NP_006793.1

View Full Product Details