Product Details

SNP ID
rs72698782
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:158578998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTATTCCTCAGGCTGAATATG[A/G]TGGGGCTGAGGAAGGGGGTAATGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10X1 PubMed Links

Gene Details

Gene
OR10X1
Gene Name
olfactory receptor family 10 subfamily X member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004477.1 902 Missense Mutation ACC,ATC T301I NP_001004477.1

View Full Product Details