Product Details

SNP ID
rs72749976
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:234630986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATTATTAAGAATCCAACAAAT[A/C]AAATATAAGTCACAGTGAATACTAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details