Product Details

SNP ID
rs73195851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126579021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTCAGTCTCCTCAAATGGAGG[A/C]CAGGACTAAGCCATGCCTCCTGGCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TXNRD3NB PubMed Links

Gene Details

Gene
TXNRD3NB
Gene Name
thioredoxin reductase 3 neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039783.1 Intron NP_001034872.1

View Full Product Details