Product Details

SNP ID
rs67343033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:182950657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTCAGAGTCTTCTACTTTTATA[C/T]ATAATTTGTTTGAAAATCAAATTCT
Phenotype
MIM: 605905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCUN1D1 PubMed Links

Gene Details

Gene
DCUN1D1
Gene Name
defective in cullin neddylation 1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308101.1 Intron NP_001295030.1
NM_020640.3 Intron NP_065691.2
XM_011512912.1 Intron XP_011511214.1
XM_011512914.1 Intron XP_011511216.1
XM_011512915.2 Intron XP_011511217.1
XM_011512916.2 Intron XP_011511218.1
XM_017006633.1 Intron XP_016862122.1
XM_017006634.1 Intron XP_016862123.1
XM_017006635.1 Intron XP_016862124.1

View Full Product Details