Product Details

SNP ID
rs73727161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151379528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGTGGCTGGAAGGGAACCAGGC[C/T]TGGGTTCAGGGGATCAGACTCTCTC
Phenotype
MIM: 607981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUB1 PubMed Links

Gene Details

Gene
NUB1
Gene Name
negative regulator of ubiquitin like proteins 1
There are no transcripts associated with this gene.

Gene
WDR86
Gene Name
WD repeat domain 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284260.1 2469 Intron NP_001271189.1
NM_001284261.1 2469 Intron NP_001271190.1
NM_001284262.1 2469 Intron NP_001271191.1
NM_198285.2 2469 Intron NP_938026.2
XM_005249989.3 2469 Intron XP_005250046.1
XM_005249990.4 2469 Intron XP_005250047.1
XM_006715966.2 2469 Intron XP_006716029.1
XM_011516144.2 2469 UTR 3 XP_011514446.1
XM_011516145.2 2469 UTR 3 XP_011514447.1
XM_011516146.2 2469 Intron XP_011514448.1
XM_011516147.2 2469 UTR 3 XP_011514449.1
XM_011516148.1 2469 Intron XP_011514450.1
XM_011516150.1 2469 Intron XP_011514452.1
XM_011516151.2 2469 UTR 3 XP_011514453.1
XM_011516152.1 2469 Intron XP_011514454.1

View Full Product Details