Product Details

SNP ID
rs72904501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:49053981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATAACTATATTGGTATCTGCTGT[C/T]CTAGAATCTCGACAGACTCCCAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM64C PubMed Links

Gene Details

Gene
TRIM64C
Gene Name
tripartite motif containing 64C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206631.1 Intron NP_001193560.1

View Full Product Details