Product Details

SNP ID
rs73955173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113169358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCACAGGTAGTTCAAATCCAAT[A/G]TTTAAAAATTAAACTCTGGGCCGGG
Phenotype
MIM: 614442
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSD4 PubMed Links

Gene Details

Gene
PSD4
Gene Name
pleckstrin and Sec7 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012455.2 Intron NP_036587.2
XM_005263634.2 Intron XP_005263691.1
XM_006712392.2 Intron XP_006712455.1
XM_006712393.2 Intron XP_006712456.1
XM_006712394.2 Intron XP_006712457.1

View Full Product Details