Product Details

SNP ID
rs72681249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77321501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCCTCCACTAAGGCTTCCAAA[C/T]GTCGCCCCAAGCCTCCCAATTTCTC
Phenotype
MIM: 603758 MIM: 607439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTZ1 PubMed Links

Gene Details

Gene
GSTZ1
Gene Name
glutathione S-transferase zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312660.1 291 Intron NP_001299589.1
NM_145870.2 291 Intron NP_665877.1
NM_145871.2 291 Intron NP_665878.2
XM_011536670.2 291 UTR 5 XP_011534972.1
XM_011536671.2 291 Intron XP_011534973.1
XM_017021243.1 291 Intron XP_016876732.1
Gene
POMT2
Gene Name
protein O-mannosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013382.5 291 Intron NP_037514.2
XM_011536675.2 291 Intron XP_011534977.1
XM_011536676.2 291 Intron XP_011534978.1
XM_011536677.2 291 Intron XP_011534979.1
XM_011536679.2 291 Intron XP_011534981.1
XM_017021244.1 291 Intron XP_016876733.1
XM_017021245.1 291 Intron XP_016876734.1

View Full Product Details