Product Details

SNP ID
rs77810585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189563782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCGGATATCTTCAAAAGGAGAAA[A/C]GGACAAGTCCAGGGGGCTTCCAGGC
Phenotype
MIM: 604653
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC40A1 PubMed Links

Gene Details

Gene
SLC40A1
Gene Name
solute carrier family 40 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014585.5 1690 Missense Mutation NP_055400.1
XM_005246505.1 1690 Missense Mutation XP_005246562.1
XM_017003938.1 1690 Missense Mutation XP_016859427.1

View Full Product Details