Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001114.4 | 5760 | UTR 3 | NP_001105.1 | ||
NM_001286057.1 | 5760 | Intron | NP_001272986.1 | ||
XM_011522835.2 | 5760 | UTR 3 | XP_011521137.1 | ||
XM_011522836.2 | 5760 | UTR 3 | XP_011521138.1 | ||
XM_011522837.2 | 5760 | UTR 3 | XP_011521139.1 | ||
XM_011522838.2 | 5760 | UTR 3 | XP_011521140.1 | ||
XM_011522839.2 | 5760 | UTR 3 | XP_011521141.1 | ||
XM_011522840.2 | 5760 | UTR 3 | XP_011521142.1 | ||
XM_011522842.2 | 5760 | UTR 3 | XP_011521144.1 | ||
XM_017022895.1 | 5760 | UTR 3 | XP_016878384.1 | ||
XM_017022896.1 | 5760 | UTR 3 | XP_016878385.1 | ||
XM_017022897.1 | 5760 | UTR 3 | XP_016878386.1 | ||
XM_017022898.1 | 5760 | Intron | XP_016878387.1 | ||
XM_017022899.1 | 5760 | Intron | XP_016878388.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001173984.2 | 5760 | Intron | NP_001167455.1 | ||
NM_013263.4 | 5760 | Intron | NP_037395.2 | ||
XM_011523046.2 | 5760 | Intron | XP_011521348.1 | ||
XM_011523047.2 | 5760 | Intron | XP_011521349.1 | ||
XM_011523048.2 | 5760 | Intron | XP_011521350.1 | ||
XM_011523049.2 | 5760 | Intron | XP_011521351.1 | ||
XM_011523050.2 | 5760 | Intron | XP_011521352.1 | ||
XM_017023179.1 | 5760 | Intron | XP_016878668.1 | ||
XM_017023180.1 | 5760 | Intron | XP_016878669.1 | ||
XM_017023181.1 | 5760 | Intron | XP_016878670.1 |