Product Details

SNP ID
rs937053
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:112703607 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GTCATTCAGTGACTGGGGGGCGGGG[A/T]CTCAGGGTGGGGTGGGAAGGGCATT
Phenotype
MIM: 610742 MIM: 609957 MIM: 165380
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MOV10 PubMed Links

Gene Details

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
There are no transcripts associated with this gene.

Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

Gene
RHOC
Gene Name
ras homolog family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042678.1 Intron NP_001036143.1
NM_001042679.1 Intron NP_001036144.1
NM_175744.4 Intron NP_786886.1

View Full Product Details