Product Details

SNP ID
hCV1350281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91272585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGCCGTGAGAGTCGGAAGGCGC[G/T]TCAGTAGTTTTCAGGTTTGCGAGCT
Phenotype
MIM: 611204 MIM: 601404
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC88C PubMed Links

Gene Details

Gene
CCDC88C
Gene Name
coiled-coil domain containing 88C
There are no transcripts associated with this gene.

Gene
GPR68
Gene Name
G protein-coupled receptor 68
There are no transcripts associated with this gene.

View Full Product Details