Product Details
- SNP ID
-
rs11781021
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:617346 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCCCCCACCATGTCCAGAGTAT[A/C]CCCTTTGCTCATGGCTCCATGCCTG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ERICH1
PubMed Links
Gene Details
- Gene
- ERICH1
- Gene Name
- glutamate rich 1
View Full Product Details