Product Details

SNP ID
rs3825011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72579094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTCCCAGGGCCCAACCCAGAG[C/T]GGTCCAGGGAATTGGGCACCCTTGA
Phenotype
MIM: 602658
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01537 PubMed Links

Gene Details

Gene
LINC01537
Gene Name
long intergenic non-protein coding RNA 1537
There are no transcripts associated with this gene.

Gene
PDE2A
Gene Name
phosphodiesterase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143839.3 Intron NP_001137311.1
NM_001146209.2 Intron NP_001139681.1
NM_001243784.1 Intron NP_001230713.1
NM_002599.4 Intron NP_002590.1
XM_005274040.2 Intron XP_005274097.1
XM_011545096.2 Intron XP_011543398.1
XM_017017910.1 Intron XP_016873399.1

View Full Product Details