Product Details

SNP ID
rs3764538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51965334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCCACATTCACTACATTCAAA[T/G]GGTTTCTCTCCTGTGTGAATTCTTT
Phenotype
MIM: 605422
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF350 PubMed Links

Gene Details

Gene
ZNF350
Gene Name
zinc finger protein 350
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021632.3 1506 Silent Mutation NP_067645.3
XM_011527196.1 1506 Silent Mutation XP_011525498.1
XM_017027094.1 1506 Silent Mutation XP_016882583.1
XM_017027095.1 1506 Silent Mutation XP_016882584.1
XM_017027096.1 1506 Silent Mutation XP_016882585.1
XM_017027097.1 1506 Silent Mutation XP_016882586.1
XM_017027098.1 1506 Silent Mutation XP_016882587.1
XM_017027099.1 1506 Silent Mutation XP_016882588.1
XM_017027100.1 1506 Silent Mutation XP_016882589.1
XM_017027101.1 1506 Silent Mutation XP_016882590.1
XM_017027102.1 1506 Silent Mutation XP_016882591.1
XM_017027103.1 1506 Silent Mutation XP_016882592.1
XM_017027104.1 1506 Silent Mutation XP_016882593.1
Gene
ZNF350-AS1
Gene Name
ZNF350 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details