Product Details
- SNP ID
-
rs35013954
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:17085829 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGGGATGGCTTTCCTCCGGGC[A/G]TGACGACAGATGAGCTGTGTGATTG
- Phenotype
-
MIM: 602129
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYO9B
PubMed Links
Gene Details
- Gene
- MYO9B
- Gene Name
- myosin IXB
View Full Product Details