Product Details

SNP ID
rs1563636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151209810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCCCCAAGACTCCAGTTCACCC[C/T]GATCACTCCTCTTCTCGGTCACCAA
Phenotype
MIM: 612510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCF2 PubMed Links

Gene Details

Gene
ABCF2
Gene Name
ATP binding cassette subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005692.4 Intron NP_005683.2
NM_007189.2 Intron NP_009120.1
XM_017011639.1 Intron XP_016867128.1
XM_017011640.1 Intron XP_016867129.1
Gene
IQCA1L
Gene Name
IQ motif containing with AAA domain 1 like
There are no transcripts associated with this gene.

View Full Product Details