Product Details

SNP ID
rs12635669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57777827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTAAACTTAAACCATGAGATGC[C/T]GTTTCTACTCAATTAAAAACTTGGA
Phenotype
MIM: 602701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLMAP PubMed Links

Gene Details

Gene
SLMAP
Gene Name
sarcolemma associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304420.2 Intron NP_001291349.1
NM_001304421.2 Intron NP_001291350.1
NM_001304422.2 Intron NP_001291351.1
NM_001304423.2 Intron NP_001291352.1
NM_001311178.1 Intron NP_001298107.1
NM_001311179.1 Intron NP_001298108.1
NM_007159.4 Intron NP_009090.2
XM_005265456.3 Intron XP_005265513.1
XM_005265457.3 Intron XP_005265514.1
XM_005265458.3 Intron XP_005265515.1
XM_005265459.3 Intron XP_005265516.1
XM_005265460.3 Intron XP_005265517.1
XM_005265461.3 Intron XP_005265518.1
XM_005265462.3 Intron XP_005265519.1
XM_005265464.3 Intron XP_005265521.1
XM_005265465.3 Intron XP_005265522.1
XM_005265466.3 Intron XP_005265523.1
XM_005265467.3 Intron XP_005265524.1
XM_005265468.3 Intron XP_005265525.1
XM_005265470.3 Intron XP_005265527.1
XM_005265471.3 Intron XP_005265528.1
XM_005265472.3 Intron XP_005265529.1
XM_006713326.2 Intron XP_006713389.1
XM_011534103.1 Intron XP_011532405.1
XM_011534104.1 Intron XP_011532406.1
XM_011534105.1 Intron XP_011532407.1
XM_011534106.1 Intron XP_011532408.1
XM_017007146.1 Intron XP_016862635.1
XM_017007147.1 Intron XP_016862636.1
XM_017007148.1 Intron XP_016862637.1
XM_017007149.1 Intron XP_016862638.1
XM_017007150.1 Intron XP_016862639.1
XM_017007151.1 Intron XP_016862640.1
XM_017007152.1 Intron XP_016862641.1
XM_017007153.1 Intron XP_016862642.1
XM_017007154.1 Intron XP_016862643.1
XM_017007155.1 Intron XP_016862644.1
XM_017007156.1 Intron XP_016862645.1
XM_017007157.1 Intron XP_016862646.1
XM_017007158.1 Intron XP_016862647.1
XM_017007159.1 Intron XP_016862648.1
XM_017007160.1 Intron XP_016862649.1
XM_017007161.1 Intron XP_016862650.1
XM_017007162.1 Intron XP_016862651.1
XM_017007163.1 Intron XP_016862652.1
XM_017007164.1 Intron XP_016862653.1
XM_017007165.1 Intron XP_016862654.1
XM_017007166.1 Intron XP_016862655.1
XM_017007167.1 Intron XP_016862656.1
XM_017007168.1 Intron XP_016862657.1
XM_017007169.1 Intron XP_016862658.1
XM_017007170.1 Intron XP_016862659.1

View Full Product Details