Product Details

SNP ID
rs7228084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50283981 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCGGCGTTCCATTTCCTGAAGGCG[A/G]GTGCGGGCACTCTGCTGCTCTCGGC
Phenotype
MIM: 609150 MIM: 156535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXXC1 PubMed Links

Gene Details

Gene
CXXC1
Gene Name
CXXC finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101654.1 1804 Silent Mutation ACC,ACT T446T NP_001095124.1
NM_014593.3 1804 Silent Mutation ACC,ACT T442T NP_055408.2
XM_011525940.2 1804 Silent Mutation ACC,ACT T446T XP_011524242.1
XM_011525941.2 1804 Silent Mutation ACC,ACT T321T XP_011524243.1
XM_017025718.1 1804 Silent Mutation ACC,ACT T442T XP_016881207.1
XM_017025719.1 1804 Silent Mutation ACC,ACT T317T XP_016881208.1
Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
There are no transcripts associated with this gene.

View Full Product Details