Product Details

SNP ID
rs12722605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:6011200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTAATCGTGTGATGTTGTTGAGC[A/T]AGAATGTTGACTCAACTGATTCGTG
Phenotype
MIM: 147730
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IL2RA PubMed Links

Gene Details

Gene
IL2RA
Gene Name
interleukin 2 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000417.2 2710 UTR 3 NP_000408.1
NM_001308242.1 2710 UTR 3 NP_001295171.1
NM_001308243.1 2710 UTR 3 NP_001295172.1

View Full Product Details