Product Details

SNP ID
rs7560440
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:237968386 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAACCAGCGTACTCCTGTGCCGAGC[A/G]GGAGGTTCCTGGGCACTGGCATTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2F PubMed Links

Gene Details

Gene
UBE2F
Gene Name
ubiquitin conjugating enzyme E2 F (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278305.1 Intron NP_001265234.1
NM_001278306.1 Intron NP_001265235.1
NM_001278307.1 Intron NP_001265236.1
NM_001278308.1 Intron NP_001265237.1
NM_080678.2 Intron NP_542409.1
Gene
UBE2F-SCLY
Gene Name
UBE2F-SCLY readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details