Product Details

SNP ID
rs131155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44495367 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTGTCCTTCTGCAATCAAGCCA[A/G]GACCCTTCAGACAGGGGCAGCTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LDOC1L PubMed Links

Gene Details

Gene
LDOC1L
Gene Name
leucine zipper down-regulated in cancer 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032287.2 2527 UTR 3 NP_115663.2

View Full Product Details