Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001029882.3 | Intron | NP_001025053.1 | |||
XM_005245848.3 | Intron | XP_005245905.1 | |||
XM_005245849.3 | Intron | XP_005245906.1 | |||
XM_005245850.3 | Intron | XP_005245907.1 | |||
XM_005245851.3 | Intron | XP_005245908.1 | |||
XM_005245852.3 | Intron | XP_005245909.1 | |||
XM_011541255.1 | Intron | XP_011539557.1 | |||
XM_011541256.2 | Intron | XP_011539558.1 | |||
XM_011541257.2 | Intron | XP_011539559.1 |