Product Details

SNP ID
rs2885057
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:57501126 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGGTCAGCCAGGCTGGTGGAGCAGC[A/G]CCTTTGGTGCCTAAGTTTCCCTAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF419 PubMed Links

Gene Details

Gene
ZNF419
Gene Name
zinc finger protein 419
There are no transcripts associated with this gene.

Gene
ZNF773
Gene Name
zinc finger protein 773
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304334.1 Intron NP_001291263.1
NM_001304335.1 Intron NP_001291264.1
NM_001304336.1 Intron NP_001291265.1
NM_001304337.1 Intron NP_001291266.1
NM_198542.2 Intron NP_940944.1

View Full Product Details