Product Details

SNP ID
rs194504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90211904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGCAGCTGGAGTGCGACCGCC[A/G]CGGCAGCCACCCTGCAACCGCCAGT
Phenotype
MIM: 605094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STEAP2 PubMed Links

Gene Details

Gene
STEAP2
Gene Name
STEAP2 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040665.1 219 UTR 5 NP_001035755.1
NM_001040666.1 219 Intron NP_001035756.1
NM_001244944.1 219 UTR 5 NP_001231873.1
NM_001244945.1 219 UTR 5 NP_001231874.1
NM_001244946.1 219 UTR 5 NP_001231875.1
NM_152999.3 219 UTR 5 NP_694544.2
XM_006715921.3 219 UTR 5 XP_006715984.1
XM_017011953.1 219 UTR 5 XP_016867442.1
XM_017011954.1 219 UTR 5 XP_016867443.1
XM_017011955.1 219 UTR 5 XP_016867444.1
XM_017011956.1 219 UTR 5 XP_016867445.1
XM_017011957.1 219 UTR 5 XP_016867446.1
XM_017011958.1 219 Intron XP_016867447.1
Gene
STEAP2-AS1
Gene Name
STEAP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details