Product Details
- SNP ID
-
rs619054
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:116790097 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGAGCCAAGGCTCCCCAGACAAG[A/G]AGCTGGGAATGGGCCTGGGCAGGTA
- Phenotype
-
MIM: 606368
MIM: 603901
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APOA5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34089864] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APOA5
- Gene Name
- apolipoprotein A5
- Gene
- ZPR1
- Gene Name
- ZPR1 zinc finger
View Full Product Details