Product Details

SNP ID
rs396275
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:109986431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCGCTGTTTTCTCACAAAATTAA[A/G]AATTTAATTCCTTTAAAAGATTGTG
Phenotype
MIM: 607826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHCYL1 PubMed Links

Gene Details

Gene
AHCYL1
Gene Name
adenosylhomocysteinase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242673.1 Intron NP_001229602.1
NM_001242674.1 Intron NP_001229603.1
NM_001242675.1 Intron NP_001229604.1
NM_001242676.1 Intron NP_001229605.1
NM_006621.5 Intron NP_006612.2
XM_011540535.1 Intron XP_011538837.1

View Full Product Details