Product Details

SNP ID
rs759848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185916322 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTTAAACAGCTATTTGAACTAAG[C/G]TAAGTTACTCTACATCATTTCATTG
Phenotype
MIM: 602719
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRA2B PubMed Links

Gene Details

Gene
TRA2B
Gene Name
transformer 2 beta homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243879.1 2536 UTR 3 NP_001230808.1
NM_004593.2 2536 UTR 3 NP_004584.1
XM_005247703.1 2536 Intron XP_005247760.1
XM_011513072.2 2536 Intron XP_011511374.1
XM_017007033.1 2536 Intron XP_016862522.1

View Full Product Details