Product Details
- SNP ID
-
rs505276
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43277240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGTTGCCACCAAGCTGAAACCC[G/T]TTTCTGTCCCCACCATCTAGTTCAA
- Phenotype
-
MIM: 606776
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TGM7
PubMed Links
Gene Details
- Gene
- TGM7
- Gene Name
- transglutaminase 7
View Full Product Details