Product Details
- SNP ID
-
rs556851
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43279021 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTGCTGGAGCCATCGGTGTGGTG[A/T]GAGGTGGGAACAGTGTCTTGTTGGC
- Phenotype
-
MIM: 606776
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TGM7
PubMed Links
Gene Details
- Gene
- TGM7
- Gene Name
- transglutaminase 7
View Full Product Details