Product Details
- SNP ID
-
rs2254667
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:92299067 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCAAACTCCTGTTCTCCACCAATC[C/G]TACCTTGGCCTCCCAAAGTGTTAGG
- Phenotype
-
MIM: 610606
MIM: 610637
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CPEB3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145168980] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPEB3
- Gene Name
- cytoplasmic polyadenylation element binding protein 3
There are no transcripts associated with this gene.
- Gene
- MARCH5
- Gene Name
- membrane associated ring-CH-type finger 5
View Full Product Details