Product Details

SNP ID
rs11911848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32778799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACAGCCTAGGAGTCCCTGATCTA[C/G]GATTTTCCTTTTTTTAAAAAAACTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C21orf62-AS1 PubMed Links

Gene Details

Gene
C21orf62-AS1
Gene Name
C21orf62 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PAXBP1
Gene Name
PAX3 and PAX7 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details