Product Details
- SNP ID
-
rs9624260
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23649989 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGAGCCTCTGTCACTCAGCATCTC[C/T]GAGCTGAGGCCAGAACAGGGGAGCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GUSBP11
PubMed Links
Gene Details
- Gene
- GUSBP11
- Gene Name
- glucuronidase, beta pseudogene 11
There are no transcripts associated with this gene.
View Full Product Details