Product Details

SNP ID
rs846899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44613413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCCATTCGTTGAGCGCCTACTAC[A/G]TGACCAGGCACTACTCCAAGCACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF23 PubMed Links

Gene Details

Gene
IGSF23
Gene Name
immunoglobulin superfamily member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205280.1 837 Intron NP_001192209.1
XM_017026317.1 837 Intron XP_016881806.1
XM_017026318.1 837 UTR 5 XP_016881807.1
XM_017026319.1 837 Intron XP_016881808.1
XM_017026320.1 837 Intron XP_016881809.1
XM_017026321.1 837 Intron XP_016881810.1
XM_017026322.1 837 Intron XP_016881811.1
XM_017026323.1 837 Intron XP_016881812.1
XM_017026324.1 837 Intron XP_016881813.1
XM_017026325.1 837 Intron XP_016881814.1
XM_017026326.1 837 Intron XP_016881815.1

View Full Product Details