Product Details
- SNP ID
-
rs2586465
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:50348606 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCACGAACCTCCAGAGAGCCACTT[G/T]ATAGTTTCCAAAGCTTCCACAAACA
- Phenotype
-
MIM: 608125
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
XYLT2
PubMed Links
Gene Details
- Gene
- XYLT2
- Gene Name
- xylosyltransferase 2
View Full Product Details