Product Details

SNP ID
rs11880364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35497892 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACCCTAAGGTGCTTTCCGAGGG[C/T]CCTAGGTCATTTTCTTTCTTCTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMKN PubMed Links

Gene Details

Gene
DMKN
Gene Name
dermokine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035516.3 Intron NP_001030593.1
NM_001126056.2 Intron NP_001119528.2
NM_001126057.2 Intron NP_001119529.2
NM_001126058.2 Intron NP_001119530.2
NM_001126059.2 Intron NP_001119531.1
NM_001190347.1 Intron NP_001177276.1
NM_001190348.1 Intron NP_001177277.1
NM_001190349.1 Intron NP_001177278.1
NM_001308380.1 Intron NP_001295309.1
NM_001308383.1 Intron NP_001295312.1
NM_033317.4 Intron NP_201574.3
XM_006723477.1 Intron XP_006723540.1
XM_006723484.1 Intron XP_006723547.1
XM_006723489.1 Intron XP_006723552.1
XM_006723493.2 Intron XP_006723556.1
XM_006723494.2 Intron XP_006723557.1
XM_006723503.2 Intron XP_006723566.1
XM_011527494.2 Intron XP_011525796.1
XM_011527495.2 Intron XP_011525797.1
XM_011527496.2 Intron XP_011525798.1
XM_011527497.2 Intron XP_011525799.1
XM_011527498.2 Intron XP_011525800.1
XM_011527499.2 Intron XP_011525801.1
XM_011527500.2 Intron XP_011525802.1
XM_011527501.2 Intron XP_011525803.1
XM_011527502.2 Intron XP_011525804.1
XM_011527503.2 Intron XP_011525805.1
XM_011527504.2 Intron XP_011525806.1
XM_011527505.2 Intron XP_011525807.1
XM_011527506.2 Intron XP_011525808.1
XM_011527507.2 Intron XP_011525809.1
XM_011527508.2 Intron XP_011525810.1
XM_011527509.2 Intron XP_011525811.1
XM_011527510.2 Intron XP_011525812.1
XM_011527511.2 Intron XP_011525813.1
XM_011527512.2 Intron XP_011525814.1
XM_011527513.1 Intron XP_011525815.1
XM_011527514.1 Intron XP_011525816.1
XM_017027475.1 Intron XP_016882964.1
XM_017027476.1 Intron XP_016882965.1
XM_017027477.1 Intron XP_016882966.1
Gene
KRTDAP
Gene Name
keratinocyte differentiation associated protein
There are no transcripts associated with this gene.

View Full Product Details