Product Details

SNP ID
rs9915115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32357839 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTGAGGGACTCTCCCTCTC[A/G]CGCAGGCTGGATTGCAGTGGTGTGA
Phenotype
MIM: 603428
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf75 PubMed Links

Gene Details

Gene
C17orf75
Gene Name
chromosome 17 open reading frame 75
There are no transcripts associated with this gene.

Gene
MIR632
Gene Name
microRNA 632
There are no transcripts associated with this gene.

Gene
ZNF207
Gene Name
zinc finger protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032293.2 Intron NP_001027464.1
NM_001098507.1 Intron NP_001091977.1
NM_003457.3 Intron NP_003448.1
XM_011525185.2 Intron XP_011523487.1
XM_017025019.1 Intron XP_016880508.1
XM_017025020.1 Intron XP_016880509.1

View Full Product Details