Product Details

SNP ID
rs797821
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:83961536 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAACTTGTTCACAGAACTCATCCAT[C/T]GTGTTGAGATTGGGGTGGTTGATGA
Phenotype
MIM: 603961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA3A PubMed Links

Gene Details

Gene
SEMA3A
Gene Name
semaphorin 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006080.2 2501 Silent Mutation ACA,ACG T717T NP_006071.1
XM_005250110.3 2501 Silent Mutation ACA,ACG T717T XP_005250167.1
XM_005250111.4 2501 Silent Mutation ACA,ACG T717T XP_005250168.1
XM_006715839.3 2501 Silent Mutation ACA,ACG T717T XP_006715902.1
XM_011515734.2 2501 Silent Mutation ACA,ACG T717T XP_011514036.1
XM_017011673.1 2501 Silent Mutation ACA,ACG T717T XP_016867162.1

View Full Product Details