Product Details

SNP ID
rs1052343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:83959826 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAAAGAAACCACAGTATAATGCT[G/T]AAAGTTTTAAATATCTCAAAGTCAG
Phenotype
MIM: 603961
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEMA3A PubMed Links

Gene Details

Gene
SEMA3A
Gene Name
semaphorin 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006080.2 4211 UTR 3 NP_006071.1
XM_005250110.3 4211 UTR 3 XP_005250167.1
XM_005250111.4 4211 UTR 3 XP_005250168.1
XM_006715839.3 4211 UTR 3 XP_006715902.1
XM_011515734.2 4211 UTR 3 XP_011514036.1
XM_017011673.1 4211 UTR 3 XP_016867162.1

View Full Product Details